Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
G6PD Polyclonal Antibody
G6PD Polyclonal Antibody
Supplier: Thermo Scientific PA518734
Description
G6PD Polyclonal Antibody for Western Blot, IHC, ELISA

Specifications
G6PD | |
Polyclonal | |
Unconjugated | |
G6PD | |
G28A; G6PD; G6PD1; G6pd-1; G6pdx; Glucose-6-phosphate 1-dehydrogenase; glucose-6-phosphate 1-dehydrogenase X; glucose-6-phosphate dehydrogenase; glucose-6-phosphate dehydrogenase X-linked; Gpdx | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
2539 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P11413 | |
G6PD | |
Synthetic peptide sequence (STNSDDVRDEKVK) corresponding to the internal amino acids of G6PD (aa 278-291). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction