Learn More
Blackline Safety™ G7c Cellular Wireless Connected Safety Device
Connected lone worker safety devices delivering leading edge technology with safety monitoring
Supplier: Blackline Safety™ G7CQHIMONA
This item is not returnable.
View return policy
Description
Specifications
Fall detection and No-motion detection; Emergency latch (Pull latch to trigger remote alert); Silent emergency (Press and hold latch to trigger remote alert); Low-battery; Worker check-in (Configurable timer: 30 - 180 min, or off, automatic check-in when driving) | |
Rechargeable Li-ion battery: 1250 mAh Li-ion | |
66 x 150 x 27 mm (2.5 x 5.9 x 1.1 in.) | |
Quad-Gas | |
-20°C to 55°C (-4°F to 131°F) | |
6.8 oz. |
18 hours continuous use at 20°C (68°F) | |
Cellular device with Quad-gas sensor cartridge with choice of: H2S, LEL, O2, CO, NH3, SO2 or Cl2 | |
IP67 | |
168 by 144 pixel graphical, high contrast, liquid crystal display with front lighting, menu system driven by a three-button keypad, power button (on/off ), check-in button (check-in/silent emergency), emergency latch (send emergency alert) Multi-language support (EN, FR, ES, NL, DE, IT) | |
2 years for G7; Lifetime for cartridges | |
192 g |