Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gab1 Rabbit anti-Human,Bovine,Mouse,Hamster,Rat, KHL, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 06579I100UG
Description
Gab1 Polyclonal antibody specifically detects Gab1 in Human,Bovine,Mouse,Hamster,Rat samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).Specifications
Gab1 | |
Polyclonal | |
NP_002030 | |
GAB1 | |
KLH-conjugated linear peptide corresponding to 31 amino acids from the C-terminal region of human GRB2-associated-binding protein 1 (Gab1). | |
Affinity Purified | |
Signaling | |
This rabbit polyclonal antibody specifically detects GRB2-associated-binding protein 1 (Gab1). It targets an epitope within 31 amino acids from the C-terminal region. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
KLH | |
GRB2-associated-binding protein 1;GRB2-associated binder 1;Growth factor receptor bound protein 2-associated protein 1 | |
Rabbit | |
∼100 kDa observed; 76.62 kDa calculated. Uncharacterized bands may be observed in some lysate(s). | |
100 μg | |
Primary | |
Human, Bovine, Mouse, Hamster, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction