Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gab1 Rabbit anti-Human,Bovine,Mouse,Hamster,Rat, KHL, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$147.00 - $372.00

Specifications

Antigen Gab1
Applications Western Blot, Immunohistochemistry (Paraffin)
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
06579I100MI
View Documents
MilliporeSigma
06579I100UG
100 μg
Each for $372.00
Add to cart
 
06579I25UMI
View Documents
MilliporeSigma
06579I25UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

Gab1 Polyclonal antibody specifically detects Gab1 in Human,Bovine,Mouse,Hamster,Rat samples. It is validated for Western Blotting, Immunohistochemistry (Paraffin).
Specifications

Specifications

Gab1
KHL
Rabbit
GRB2-associated-binding protein 1;GRB2-associated binder 1;Growth factor receptor bound protein 2-associated protein 1
KLH-conjugated linear peptide corresponding to 31 amino acids from the C-terminal region of human GRB2-associated-binding protein 1 (Gab1).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Western Blot, Immunohistochemistry (Paraffin)
Purified
Human, Bovine, Mouse, Hamster, Rat
GAB1
Polyclonal
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit