Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAB1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GAB1
Applications Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095945
View Documents
Novus Biologicals
NBP304453100UL
100 μg
Each for $320.71
Add to cart
 
NB095946
View Documents
Novus Biologicals
NBP30445320UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

GAB1 Polyclonal antibody specifically detects GAB1 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

GAB1
Unconjugated
Rabbit
Apoptosis, Phospho Specific, Signal Transduction, Tyrosine Kinases
PBS with 50% glycerol, pH7.3.
2549
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunohistochemistry
Purified
RUO
Human
GRB2-associated binder 1, GRB2-associated binding protein 1, GRB2-associated-binding protein 1, Growth factor receptor bound protein 2-associated protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human GAB1 (NP_997006.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit