Learn More
GAB2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 225491AP
Catalog No. 50-173-3885
Add to cart
Description
Description
This gene is a member of the GRB2-associated binding protein (GAB) gene family and is similar to the GAB1 gene. These proteins contain pleckstrin homology (PH) domain, and bind SHP2 tyrosine phosphatase and GRB2 adapter protein. They act as adapters for transmitting various signals in response to stimuli through cytokine and growth factor receptors, and T- and B-cell antigen receptors. The protein encoded by this gene is the principal activator of phosphatidylinositol-3 kinase in response to activation of the high affinity IgE receptor. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
Specifications
Specifications
GAB2
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UQC2, Q9Z1S8
GAB2, GRB2 associated binder 2, KIAA0571, pp100
Rabbit
IgG
150 μL
-20°C
Primary
14389, 9846
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GAB2
Liquid
GAB2
GAB2 Fusion Protein Ag18192
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse