Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAB4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548225

Catalog No. PA548225

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Specifications

Specifications

GAB4
Polyclonal
PBS with 0.09% sodium azide
Q2WGN9
GRB2-associated-binding protein 4, GRB2-associated binder 2-like, GAB2-like, GRB2-associated binder 4, GRB2-associated-binding protein 2-like, Growth factor receptor bound protein 2-associated protein 4, GAB4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
128954
Flow Cytometry, Western Blot
Unconjugated
GAB4
Liquid
GAB4
KLH conjugated synthetic peptide between 64-92 amino acids from the N-terminal region of human GAB4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit