Learn More
GABA-AR alpha 2 Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: S399-19, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259325G
Catalog No. NBP259325G
Add to cart
Description
Description
GABA-AR alpha 2 Monoclonal antibody specifically detects GABA-AR alpha 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
GABA-AR alpha 2
S399-19
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
GABRA2
Fusion protein amino acids 350-385 (Cytoplasmic C-terminus) of rat GABA-A Receptor Alpha2
Protein G purified
Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
2555
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
GABA(A) receptor subunit alpha-2, gamma-aminobutyric acid (GABA) A receptor, alpha 2, gamma-aminobutyric acid receptor subunit alpha-2
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Detects 55kDa. Does not cross-react with GABA-A Receptor Alpha1.