Learn More
GABA-AR alpha 4 Mouse anti-Human, Mouse, Rat, Alexa Fluor 405, Clone: S398A-34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP259326AF405
Catalog No. NB259326405
Add to cart
Description
Description
GABA-AR alpha 4 Monoclonal specifically detects GABA-AR alpha 4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
GABA-AR alpha 4
S398A-34
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
GABRA4
Fusion protein amino acids 410-450 (Cytoplasmic C-terminus) of rat GABA-A-Receptor Alpha 4
Protein G purified
Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary
2557
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
GABA(A) receptor subunit alpha-4, gamma-aminobutyric acid (GABA) A receptor, alpha 4, gamma-aminobutyric acid receptor subunit alpha-4
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Detects 60kDa. Does not cross-react with other GABA-A-Receptor Alpha subunits.