Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 5 Mouse anti-Human, Alexa Fluor 647, Clone: OTI1G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270789AF647

Catalog No. N270789A647


Add to cart

Description

Description

GABA-AR alpha 5 Monoclonal antibody specifically detects GABA-AR alpha 5 in Human samples. It is validated for Western Blot.
Specifications

Specifications

GABA-AR alpha 5
OTI1G9
Western Blot
Affinity Purified, Conjugated
GABRA5
Recombinant protein fragment corresponding to amino acids 32-259 of human GABA-A R alpha 5 (NP_000801) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184
Mouse
IgG2b
0.1 mL
Primary
2558.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit