Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-A R beta 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | GABA-A R beta 1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GABA-A R beta 1 Polyclonal specifically detects GABA-A R beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
GABA-A R beta 1 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
GABA(A) receptor subunit beta-1, gamma-aminobutyric acid (GABA) A receptor, beta 1, gamma-aminobutyric acid receptor subunit beta-1 | |
A synthetic peptide of human GABA-A R beta 1. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS with 50% glycerol, pH7.3. | |
2560 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title