Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-A R beta 2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GABA-A R beta 2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094808
View Documents
Novus Biologicals
NBP303626100UL
100 μg
Each for $320.71
Add to cart
 
NB094809
View Documents
Novus Biologicals
NBP30362620UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

GABA-A R beta 2 Polyclonal antibody specifically detects GABA-A R beta 2 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

GABA-A R beta 2
Unconjugated
Rabbit
Neuronal Cell Markers, Neuroscience, Neurotransmission
PBS with 50% glycerol, pH7.3.
2561
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse
GABA(A) receptor subunit beta-2, gamma-aminobutyric acid (GABA) A receptor, beta 2, gamma-aminobutyric acid A receptor beta 2, gamma-aminobutyric acid receptor subunit beta-2, MGC119386, MGC119388, MGC119389
Recombinant fusion protein containing a sequence corresponding to amino acids 26-244 of human GABA-A R beta 2 (NP_000804.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit