Learn More
GABA-A R beta 3 Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: S87-25, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP147613C
Catalog No. NBP147613C
Add to cart
Description
Description
GABA-A R beta 3 Monoclonal specifically detects GABA-A R beta 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
GABA-A R beta 3
S87-25
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
P63080
ECA5, GABA(A) receptor beta-3 subunit, GABA(A) receptor subunit beta-3, GABAA receptor beta-3 subunit, GABA-alpha receptor beta-2 subunit, gamma-aminobutyric acid (GABA) A receptor, beta 3, gamma-aminobutyric acid receptor subunit beta-3, MGC9051
Mouse
IgG1
0.1 ml
Primary
2562
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Purified
GABRB3
Fusion protein amino acids 370-433 of mouse GABA-A-R-Beta 3
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 55kDa. No cross-reactivity against GABA-A-R-Beta 2 or -Beta1.