Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA-A R gamma 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 12 publications
Supplier: Novus Biologicals NB300190
Description
GABA-A R gamma 2 Polyclonal specifically detects GABA-A R gamma 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Knockdown Validated.Specifications
GABA-A R gamma 2 | |
Polyclonal | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
CAE2, ECA2, GABA(A) receptor subunit gamma-2, gamma-aminobutyric acid (GABA) A receptor, gamma 2, gamma-aminobutyric acid receptor subunit gamma-2, GEFSP3 | |
Rabbit | |
46 kDa | |
0.1 mL | |
Neuronal Cell Markers, Neuroscience, Neurotransmission | |
2566 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, KnockDown | |
0.15 mg/mL | |
Western Blot 1:1000, Immunohistochemistry 1:100, KnockDown Validated | |
P18508 | |
GABRG2 | |
Fusion protein from the cytoplasmic loop of the gamma 2 subunit. Accession # P18508 | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Specific for the ∼46k gamma2-subunit of the GABAA receptor in Western blots. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction