Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GABA-AR alpha 2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093556
View Documents
Novus Biologicals
NBP302985100UL
100 μg Each for $320.71
Add to cart
 
NB093557
View Documents
Novus Biologicals
NBP30298520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

GABA-AR alpha 2 Polyclonal antibody specifically detects GABA-AR alpha 2 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

GABA-AR alpha 2
Unconjugated
Rabbit
Neuronal Cell Markers, Neuroscience, Neurotransmission
PBS with 50% glycerol, pH7.3.
2555
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse
GABA(A) receptor subunit alpha-2, gamma-aminobutyric acid (GABA) A receptor, alpha 2, gamma-aminobutyric acid receptor subunit alpha-2
A synthetic peptide corresponding to a sequence within amino acids 300-400 of human GABA-AR alpha 2 (NP_000798.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit