Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABA-AR alpha 5 Antibody (OTI1G9), Biotin, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP270789B

Catalog No. NBP270789B


Add to Cart

Description

Description

GABA-AR alpha 5 Monoclonal specifically detects GABA-AR alpha 5 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

GABA-AR alpha 5
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
GABRA5
Recombinant protein fragment corresponding to amino acids 32-259 of human GABA-A R alpha 5 (NP_000801) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG2b
Western Blot
OTI1G9
Western Blot
GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184
Mouse
Affinity Purified
Cellular Markers, Neuronal Cell Markers, Neurotransmission
2558.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit