Learn More
GABA-B R1 Mouse anti-Human, Mouse, Rat, FITC, Clone: S93A-49, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259336F
Catalog No. NBP259336F
Add to cart
Description
Description
GABA-B R1 Monoclonal antibody specifically detects GABA-B R1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
GABA-B R1
S93A-49
Immunocytochemistry/Immunofluorescence
Purified
GABBR1
Fusion protein amino acids 873-977 (cytoplasmic C-terminus) of rat GABA(B)R1
Protein G purified
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary
2550
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
FLJ92613, GABABR1, GABBR1-3, gamma-aminobutyric acid (GABA) B receptor, 1, gamma-aminobutyric acid type B receptor subunit 1, GB1, GPRC3Asubunit 1c, hGB1a, seven transmembrane helix receptor
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Detects 115kDa. No cross-reactivity against GABA(B)R2.