Learn More
GABA-B R2 Mouse anti-Human, DyLight 488, Clone: 5G7B9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237280G
Catalog No. NBP237280G
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
GABA-B R2
5G7B9
Western Blot, ELISA, Immunohistochemistry
Purified
GABBR2
Purified recombinant fragment of human GABBR2 (AA: 319-483) expressed in E. Coli.
Protein A or G purified
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary
9568
ELISA, Immunohistochemistry, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
G protein-coupled receptor 51, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, Gb2, GPR51GABA-B receptor, R2 subunit, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099
Mouse
IgG2a
0.1 ml
Store at 4C in the dark.
Monoclonal
Human