Learn More
GABA-B R2 Mouse anti-Human, Mouse, Rat, DyLight 350, Clone: S81-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259335UV
Catalog No. NBP259335UV
Add to cart
Description
Description
GABA-B R2 Monoclonal antibody specifically detects GABA-B R2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
GABA-B R2
S81-2
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
GABBR2
Fusion protein amino acids 861-912 of rat GABA(B)R2
Protein G purified
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary
9568
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
G protein-coupled receptor 51, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, Gb2, GPR51GABA-B receptor, R2 subunit, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099
Mouse
IgG1
100 ul
Store at 4C in the dark.
Monoclonal
Detects 105kDa. No cross-reactivity against GABA(B)R1.