Learn More
GABA Receptor Epsilon Mouse anti-Human, Rat, DyLight 488, Clone: S413-67, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259323G
Catalog No. NBP259323G
Add to cart
Description
Description
GABA Receptor Epsilon Monoclonal antibody specifically detects GABA Receptor Epsilon in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
GABA Receptor Epsilon
S413-67
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
GABRE
Fusion protein amino acids 27-253 (Extracellular N-terminus) of human GABA A Receptor Epsilon
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 60kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
GABA(A) receptor subunit epsilon, gamma-aminobutyric acid (GABA) A receptor, epsilon, gamma-aminobutyric acid receptor subunit epsilon
Mouse
IgG2a
100 ul
Primary
2564
Human, Rat