Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABA Transporter 2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061098
Description
GABA Transporter 2 Polyclonal specifically detects GABA Transporter 2 in Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
GABA Transporter 2 | |
Polyclonal | |
Western Blot, Immunohistochemistry 1:100-1:200, Immunohistochemistry-Paraffin 1:10-1:500 | |
P31646 | |
SLC6A13 | |
Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH. Accession # P31646 | |
Affinity purified | |
RUO | |
6540 | |
Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
GABA transport protein, GAT-2, GAT2Solute carrier family 6 member 13, sodium- and chloride-dependent GABA transporter 2, solute carrier family 6 (neurotransmitter transporter, GABA), member 13 | |
Rabbit | |
82 kDa | |
0.1 mL | |
Primary | |
Specific for the ∼82k GAT-2 protein. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction