Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabapentin, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

60142-96-3
C9H17NO2
171.24
UGJMXCAKCUNAIE-UHFFFAOYSA-N
1-(Aminomethyl)cyclohexaneacetic acid
3446
CHEBI:42797

Anticonvulsant. Increases brain GABA

Anticonvulsant with several possible mechanisms of action. Increases GABA in the brain and binds to a novel site associated with voltage-sensitive Ca2+ channels. Prevents neuronal death and is antinociceptive and anxiolytic.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
SMILES C1CCC(CC1)(CC(=O)O)CN
IUPAC Name 2-[1-(aminomethyl)cyclohexyl]acetic acid
Formula Weight 171.24
Chemical Name or Material Gabapentin
Grade >99%

Recommended Storage : Desiccate at +4°C