Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabapentin, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
60142-96-3
C9H17NO2
171.24
UGJMXCAKCUNAIE-UHFFFAOYSA-N
2-[1-(aminomethyl)cyclohexyl]acetic acid
NCC1(CC(O)=O)CCCCC1
60142-96-3
C9H17NO2
171.24
UGJMXCAKCUNAIE-UHFFFAOYSA-N
2-[1-(aminomethyl)cyclohexyl]acetic acid
NCC1(CC(O)=O)CCCCC1

G1092, 60142-96-3, C9H17NO2

Spectrum™ Gabapentin, USP is used with other medications to prevent and control seizures and to relieve nerve pain following shingles in adults. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Ignition Residue 0.001
Quantity 1 kg
CAS Max % 1
Percent Purity 98 to 102%