Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABARAPL3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549016
Description
Ubiquitin-like modifier involved in autophagosome formation. Whereas LC3s are involved in elongation of the phagophore membrane, the GABARAP/GATE-16 subfamily is essential for a later stage in autophagosome maturation.Specifications
GABARAPL3 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9BY60 | |
GABARAPL3 | |
KLH conjugated synthetic peptide between 27-53 amino acids from the Central region of human GABARAPL3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GABARAPL3 | |
Gamma-aminobutyric acid receptor-associated protein-like 3, GABA(A) receptor-associated protein-like 3, GABARAPL3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23766 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction