Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gabbr2 Mouse anti-Human, Mouse, Rat, Clone:S81-2, Abnova™
Mouse monoclonal antibody raised against recombinant rat Gabbr2.
Supplier: Abnova Corporation MAB17649
Specifications
Gabbr2 | |
Monoclonal | |
Unconjugated | |
Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
Gabbr2 | |
Mouse | |
100 μg | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
S81-2 | |
gamma-aminobutyric acid (GABA) B receptor 2 | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
Gpr51 | |
Fusion protein amino acids 861-912 of rat Gabbr2. | |
83633 | |
Store at -20°C. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction