Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gabexate Mesylate 97.0+%, TCI America™

Click to view available options
Quantity:
1g
5g
56974-61-9
C17H27N3O7S
417.477
MFCD00210299
DNTNDFLIKUKKOC-UHFFFAOYSA-N
gabexate mesylate,gabexate mesilate,gabexate monomethanesulfonate,megacart,gabexate methanesulfonate,foy,dsstox_cid_25591,dsstox_rid_80986,dsstox_gsid_45591
6604561
ethyl 4-[6-(diaminomethylideneamino)hexanoyloxy]benzoate;methanesulfonic acid
CCOC(=O)C1=CC=C(C=C1)OC(=O)CCCCCN=C(N)N.CS(=O)(=O)O
56974-61-9
C17H27N3O7S
417.477
MFCD00210299
DNTNDFLIKUKKOC-UHFFFAOYSA-N
gabexate mesylate,gabexate mesilate,gabexate monomethanesulfonate,megacart,gabexate methanesulfonate,foy,dsstox_cid_25591,dsstox_rid_80986,dsstox_gsid_45591
6604561
ethyl 4-[6-(diaminomethylideneamino)hexanoyloxy]benzoate;methanesulfonic acid
CCOC(=O)C1=CC=C(C=C1)OC(=O)CCCCCN=C(N)N.CS(=O)(=O)O
Quantity 1g
Color White
Melting Point 91°C
Formula Weight 417.48
Physical Form Crystal-Powder at 20°C
Percent Purity ≥97.0% (N)
Chemical Name or Material Gabexate Mesylate

RTECSNumber : DG2800800

TSCA : No