Learn More
Thermo Scientific™ Gable Covers for Lindberg/Blue M™ Baths Available on GSA/VA Contract for Federal Government customers only.
Prevent cross-contamination by diverting condensation to sides for return to bath medium.
Manufacturer:  Thermo Scientific™ 38576G01
 View more versions of this product
Catalog No. 15-453-203A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Prevent cross-contamination by diverting condensation to sides for return to bath medium with Thermo Scientific™ Gable Bath Covers.
  • Reduce moisture loss and water accumulation on the bench
  • Stainless steel or semi-transparent polycarbonate
  • Can be attached to side of bath
  • With handle
Description & Specifications
Specifications
Model WB1110
Water Bath Models WB1110
Stainless steel
Gable Cover