Learn More
Thermo Scientific™ Gable Cover for VersaCool™ Refrigerated Circulating Bath
Reversible hinged cover for water, water/glycol applications to be used with Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Manufacturer:  Thermo Scientific™ 230000003
Catalog No. 13-275-108
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Utilize the gable cover to accommodate larger size beakers for optimal performance with the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath.
Optional accessory for the Thermo Scientific™ VersaCool™ Refrigerated Circulating Bath. This reversible hinged cover allows for larger size beakers and:
  • Reduces moisture and directs condensate back into bath
  • Ideal for water, water/glycol applications
Specifications
Specifications
Gable Cover
Gable Cover-reversible hinged cover for water, water/glycol applications
Thermo Scientific VersaCool Refrigerated Circulating Bath