Thermo Scientific™ Gable Covers for Lindberg/Blue M™ Baths Available on GSA/VA Contract for Federal Government customers only.
Prevent cross-contamination by diverting condensation to sides for return to bath medium.
Manufacturer: Thermo Scientific™ 118107
 View more versions of this product
Catalog No. 15-453-202
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Equipment) Water Bath Models WB1110
    Type Gable Cover
    Material Polycarbonate
    For Use With Model WB1110
    Prevent cross-contamination by diverting condensation to sides for return to bath medium with Thermo Scientific™ Gable Bath Covers.
    • Reduce moisture loss and water accumulation on the bench
    • Stainless steel or semi-transparent polycarbonate
    • Can be attached to side of bath
    • With handle
  • Description & Specifications
Related Products
Product Content Feedback