Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABPB1 Mouse anti-Human, Clone: OTI7A7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527434

Catalog No. PIMA527434


Add to cart

Description

Description

Transcription factor capable of interacting with purine rich repeats (GA repeats). Necessary for the expression of the Adenovirus E4 gene.
Specifications

Specifications

GABPB1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q06547
BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4TF1B; GABPB; GABPB2; NRF2B1; NRF2B2; GA-binding protein subunit beta-1; GABP subunit beta-2; GABPB-1; nuclear respiratory factor 2; transcription factor E4TF1-47; transcription factor E4TF1-53
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2553
Immunohistochemistry (Paraffin), Western Blot
OTI7A7
Unconjugated
GABPB1
Liquid
GABPB1
Human recombit protein fragment corresponding to amino acids 128-360 of GABPB1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit