Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABRA2 Mouse anti-Human, Mouse, Rat, Clone: S399-19, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527702

Catalog No. PIMA527702


Add to cart

Description

Description

GABA is the major inhibitory neurotransmitter in the mammalian brain where it acts at GABA-A receptors, which are ligand-gated chloride channels. Chloride conductance of these channels can be modulated by agents such as benzodiazepines that bind to the GABA-A receptor. At least 16 distinct subunits of GABA-A receptors have been identified. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications

Specifications

GABRA2
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
P23576, P26048, P47869
GABA A receptor subunit alpha 2, GABRA2, Gamma aminobutyric acid A receptor Alpha 2, GBRA2_Human
Mouse
IgG1
100 μg
-20°C
Primary
14395, 2555, 289606
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S399-19
Unconjugated
GABRA2
Liquid
Gabra2
Fusion protein amino acids 350-385 (Cytoplasmic C-terminus) of rat GABA-A Receptor Alpha2
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit