Learn More
GABRB1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group Inc 201831AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-2754
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel.
Specifications
Specifications
GABRB1
Polyclonal
Unconjugated
GABRB1
GABA A receptor beta 1, GABA A receptor beta, GABAA receptor beta 1, GABRB1
Rabbit
Antigen Affinity Chromatography
RUO
14400, 25450, 2560
-20°C
Liquid
Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15431, P18505, P50571
Gabrb1
GABRB1 Fusion Protein Ag14070
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG