Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABRD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5119681

This item is not returnable.
View return policy
Description
Positive Control: HT-29 cell lysates, MCF-7. Subcellular Location: Postsynaptic cell membrane, cell membrane.
GABRD Polyclonal antibody specifically detects GABRD in Human, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
GABRD | |
Polyclonal | |
Unconjugated | |
GABRD | |
AI853201; EIG10; EJM7; GABA(A) receptor subunit delta; GABA(A) receptor, delta; GABAA receptor delta subunit; GABA-A receptor delta-subunit; GABA-A receptor, delta polypeptide; GABAA-RD; GABAARdelta; Gabrd; gamma-aminobutyric acid; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, delta; gamma-aminobutyric acid (GABA) A receptor, subunit delta; gamma-aminobutyric acid (GABA-A) receptor, subunit delta; gamma-aminobutyric acid A receptor, delta; gamma-aminobutyric acid receptor subunit delta; gamma-aminobutyric acid type A receptor delta subunit; GEFSP5; MGC105467; MGC45284 | |
Synthetic peptide within rat GABRD aa 1-80 | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
O14764, P18506 | |
Rabbit | |
Affinity chromatography | |
RUO | |
2563, 29689 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title