Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABRD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 156231AP

Catalog No. 50-173-0359


Add to cart

Description

Description

GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels, however, they are structurally and functionally distinct. Members of the GABAA receptor family include GABAA Ralpha1-6, GABAA R beta1-3, GABAA Rgamma1-3, GABAA Rdelta, GABAA Repsilon, GABAA Rrho1 and GABAA Rrho2. The GABAB family is composed of GABAB R1alpha and GABAB R1beta. GABA transporters have also been identified and include GABA T-1, GABA T-2 and GABA T-3 (also designated GAT-1, -2, and -3). The GABA transporters function to terminate GABA action.
Specifications

Specifications

GABRD
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14764, P18506, P22933
GABAA receptor delta, GABA A receptor delta, GABA(A) receptor subunit delta, GABRD
Rabbit
IgG
150 μL
-20°C
Primary
14403, 2563, 29689
Western Blot
Unconjugated
GABRD
Liquid
Gabrd
GABRD Fusion Protein Ag8064
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit