Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GABRP Polyclonal Antibody, Invitrogen™

Catalog No. PIPA124281
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA124281 100 μg
1 options

Catalog No. PIPA124281

Supplier: Thermo Scientific PA124281

Rabbit Polyclonal Antibody

PA1-24281 detects GABRP in human samples. PA1-24281 has been successfully used in Western blot procedures. PA1-24281 immunogen is a synthetic peptide corresponding to a region of human GABRP. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the gaba/benzodiazepine receptor and opening an integral chloride channel. in the uterus, the function of the receptor appears to be related to tissue contractility. The binding of this pi subunit with other GABA (A) receptor subunits alters the sensitivity of recombit receptors to modulatory agents such as pregolone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GABRP
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene GABRP
Gene Accession No. O00591
Gene Alias MGC126386, MGC126387
Gene Symbols GABRP
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region of human GABRP
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2568
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.