Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABRP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5107132
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
GABRP Polyclonal antibody specifically detects GABRP in Mouse samples. It is validated for Western Blot.Specifications
GABRP | |
Polyclonal | |
Unconjugated | |
GABRP | |
GABA(A) receptor subunit pi; GABA(A) receptor, pi; GABA-A receptor pi subunit; GABRP; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, pi; gamma-aminobutyric acid (GABA-A) receptor, pi; gamma-aminobutyric acid A receptor, pi; gamma-aminobutyric acid receptor subunit pi; gamma-aminobutyric acid type A receptor pi subunit; gamma-aminobutyric acid type A receptor subunit pi; MGC126386; MGC126387 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
216643 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
Q8QZW7 | |
GABRP | |
Synthetic peptide located within the following region: GFENLTAGYN KFLRPNFGGD PVRIALTLDI ASISSISESN MDYTATIYLR | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction