Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GABRR1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549210
Description
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel. Rho-1 GABA receptor could play a role in retinal neurotransmission.Specifications
GABRR1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P24046 | |
GABRR1 | |
KLH conjugated synthetic peptide between 19-48 amino acids from the N-terminal region of human GABRR1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GABRR1 | |
Gamma-aminobutyric acid receptor subunit rho-1, GABA(A) receptor subunit rho-1, GABA(C) receptor, GABRR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2569 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction