Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$395.30

Specifications

Antigen GAC
Concentration 0.29 mg/mL
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-553-514
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
199581AP150UL
150 μL
Each of 1 for $395.30
Add to cart
 
Description

Description

This antibody is specific to GAC isoform. It has no cross reaction to KGA.

Sahai (1983) demonstrated phosphate-activated glutaminase (EC 3.5.1.2) in human platelets. It is the major enzyme yielding glutamate from glutamine. Significance of the enzyme derives from its possible implication in behavior disturbances in which glutamate acts as a neurotransmitter(Prusiner, 1981). High heritability of platelet glutaminase was indicated by studies of Sahai and Vogel (1983) who found an intraclass correlation coefficient of 0.96 for monozygotic twins and 0.53 for dizygotic twins.
Specifications

Specifications

GAC
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
IgG
-20°C
0.29 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit