Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD1/GAD67 Mouse anti-Human, Clone: GAD1/2391, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen GAD1/GAD67
Clone GAD1/2391
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122188
View Documents
Novus Biologicals
NBP30735720UG
20 μg
Each for $138.37
Add to cart
 
NB122187
View Documents
Novus Biologicals
NBP30735710UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

GAD1/GAD67 Monoclonal antibody specifically detects GAD1/GAD67 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin), Protein Array
Specifications

Specifications

GAD1/GAD67
0.2 mg/ml
Western Blot, ELISA, Immunohistochemistry (Paraffin), Peptide Array
Purified
RUO
Human
67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
GAD1/2391
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Unconjugated
Mouse
Diabetes Research
10 mM PBS with 0.05% BSA
2571
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit