Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD1/GAD67 Mouse anti-Human, Monkey, Janelia Fluor™ 549, Clone: OTI3G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270791JF549

Catalog No. N270791J549


Add to cart

Description

Description

GAD1/GAD67 Monoclonal antibody specifically detects GAD1/GAD67 in Human, Monkey samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

GAD1/GAD67
OTI3G9
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
GAD1
Full length recombinant protein of human GAD1/GAD67 (NP_000808) produced in HEK293T cells.
Immunogen affinity purified
Diabetes Research
Primary
2571.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Monkey
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit