Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAD1/GAD67, Mouse anti-Human, PerCP, Clone: GAD1/2563, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279937PCP
Description
GAD1/GAD67 Monoclonal antibody specifically detects GAD1/GAD67 in Human samples. It is validated for Flow Cytometry.Specifications
GAD1/GAD67 | |
GAD1/2563 | |
Flow Cytometry | |
Conjugated, Purified | |
GAD1 | |
Recombinant human GAD1/GAD67 protein fragment (around aa 72-135) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Flow Cytometry | |
PerCP | |
PBS with 0.05% Sodium Azide | |
67kD), EC 4.1.1, FLJ45882, GAD25, glutamate decarboxylase 1 (brain, 67kDa), Glutamate decarboxylase 67 kDa isoform | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Diabetes Research | |
Primary | |
This monoclonal antibody recognizes a protein of 67kDa, which is identified as glutamic acid decarboxylase 1 (GAD1/GAD67). |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title