Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAD2/GAD65, Mouse anti-Human, Clone: GAD2/1960, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP27980420UG
Description
GAD2/GAD65 Monoclonal specifically detects GAD2/GAD65 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
GAD2/GAD65 | |
Monoclonal | |
0.2 mg/mL | |
Immunohistochemistry 0.1-0.2 ug/ml, Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array | |
Q05329, Q05329 | |
GAD2 | |
Recombinant human GAD2/GAD65 protein fragment (around aa 6-99) (exact sequence is proprietary) | |
20 μg | |
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience | |
2572 | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
GAD2/1960 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
EC 4.1.1, EC 4.1.1.15, GAD-65, GAD65MGC161605, glutamate decarboxylase 2, glutamate decarboxylase 2 (pancreatic islets and brain, 65kD), glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa), Glutamate decarboxylase 65 kDa isoform, Glutamate decarboxylase-2 (pancreas), MGC161607,65 kDa glutamic acid decarboxylase | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
This monoclonal antibody recognizes a protein of 65kDa, which is identified as glutamic acid decarboxylase 2 (GAD2/GAD65). | |
Store at 4°C. | |
IgG2b κ |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction