Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD2/GAD65, Mouse anti-Human, Clone: GAD2/1960, Novus Biologicals™

Mouse Monoclonal Antibody

$156.50 - $345.50

Specifications

Antigen GAD2/GAD65
Clone GAD2/1960
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB010625
View Documents
Novus Biologicals
NBP279804100UG
100 ug
Each for $345.50
Add to cart
 
NB010626
View Documents
Novus Biologicals
NBP27980420UG
20 ug
Each for $156.50
Add to cart
 
Description

Description

GAD2/GAD65 Monoclonal antibody specifically detects GAD2/GAD65 in Human Samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array.
Specifications

Specifications

GAD2/GAD65
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Human
Primary
GAD2/1960
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit