Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAD2/GAD65 Mouse anti-Human, Clone: GAD2/2362, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | GAD2/GAD65 |
---|---|
Clone | GAD2/2362 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array |
Applications | Immunohistochemistry (Paraffin), Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB123120
|
Novus Biologicals
NBP30782320UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
GAD2/GAD65 Monoclonal specifically detects GAD2/GAD65 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
GAD2/GAD65 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience | |
10 mM PBS with 0.05% BSA | |
2572 | |
IgG2b κ | |
Protein A or G purified |
GAD2/2362 | |
Immunohistochemistry-Paraffin 0.1-0.2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 4.1.1, EC 4.1.1.15, GAD-65, GAD65MGC161605, glutamate decarboxylase 2, glutamate decarboxylase 2 (pancreatic islets and brain, 65kD), glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa), Glutamate decarboxylase 65 kDa isoform, Glutamate decarboxylase-2 (pancreas), MGC161607,65 kDa glutamic acid decarboxylase | |
Recombinant fragment of human GAD2/GAD65 protein (around aa 6-99) (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title