Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GAD2/GAD65, Mouse anti-Human, Janelia Fluor 646, Clone: GAD2/1960, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279938JF646
Description
GAD2/GAD65 Monoclonal antibody specifically detects GAD2/GAD65 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
GAD2/GAD65 | |
GAD2/1960 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
GAD2 | |
Recombinant human GAD2/GAD65 protein fragment (around aa 6-99) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EC 4.1.1, EC 4.1.1.15, GAD-65, GAD65MGC161605, glutamate decarboxylase 2, glutamate decarboxylase 2 (pancreatic islets and brain, 65kD), glutamate decarboxylase 2 (pancreatic islets and brain, 65kDa), Glutamate decarboxylase 65 kDa isoform, Glutamate decarboxylase-2 (pancreas), MGC161607,65 kDa glutamic acid decarboxylase | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience | |
Primary | |
This monoclonal antibody recognizes a protein of 65kDa, which is identified as glutamic acid decarboxylase 2 (GAD2/GAD65). |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title