Sino Biological™ GAD2 (GAD65) Recombinant Mouse Protein, N-His.GST Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding full length of mouse GAD2 (Met 1-Leu 585) fused to N-terminal polyhistidine-tagged GST tag at N-terminus

Manufacturer: Sino Biological™ 50653M20B5

 View more versions of this product

Catalog No. 50-653-M20B5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Quantity 5μg
    Structural Form Full Length, Recombinant
    Protein Tag GST Tag, His Tag
    Molecular Weight 93kd
    Purity ≥90% by SDS-PAGE
    Source Mouse
    Endotoxin Concentration <1 EU/μg
    Common Name Glutamate Decarboxylase 2 (pancreatic islets and brain 65kDa)
    Gene Symbol GAD2
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Insect
    Product Type GAD2 (GAD65) Recombinant Mouse Protein
    For Use With (Application) Analysis of protein stucture and used in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 14417

    Expressed from DNA sequence encoding full length of mouse GAD2 (Met 1-Leu 585) fused to N-terminal polyhistidine-tagged GST tag at N-terminus

    • Formulation: 50mM Tris, 100mM NaCl, 0.5mM PMSF, 0.5mM GSH, 3mM DTT, 10% glycerol, pH 8.0

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit