Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GAD67 Mouse anti-Human, Mouse, Clone: GT12212, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527816

Catalog No. PIMA527816


Add to cart

Description

Description

Positive Control: mouse brain Predicted Reactivity: Human (100%), Rat (96%), Dog (99%), Cat (99%), Pig (99%), Chicken (90%), Chimpanzee (100%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

GAD-65 and GAD-67 are members of the group II decarboxylase family of proteins and are responsible for catalyzing the rate limiting step in the production of GABA ( gamma-aminobutyric acid) from L-glutamic acid. Although both GADs are found in the brain, GAD-65 localizes to synaptic vesicle membranes in nerve terminals, while GAD-67 is distributed throughout the cell. GAD-67 is responsible for the basal levels of GABA synthesis. In the case of a heightened demand for GABA in neurotransmission, GAD-65 will transiently activate to assist in GABA production. The loss of GAD-65 is detrimental and can impair GABA neurotransmission, however the loss of GAD-67 is lethal. Due to alternative splicing, two isoforms exist for GAD-67: the predomit GAD-67 form and the minor GAD-25 form.
Specifications

Specifications

GAD67
Monoclonal
1 mg/ml
PBS with no preservative
P48318, Q99259
CPSQ1, GAD, GAD1, GAD67, SCP, glutamate decarboxylase 1
Mouse
IgG2b
100 μl
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
14415, 2571
Western Blot
GT12212
Unconjugated
GAD1
Liquid
Gad1
Recombit protein encompassing a sequence within the N-terminus region of human GAD67. The exact sequence is proprietary.
Protein A
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit