Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GADD34 Antibody (OTI3D12), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271765AF647
Description
GADD34 Monoclonal specifically detects GADD34 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
GADD34 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PPP1R15A | |
Full length recombinant protein of human GADD34 (NP_055145) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG2b |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3D12 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
GADD34Growth arrest and DNA damage-inducible protein GADD34, growth arrest and DNA-damage-inducible 34, Myeloid differentiation primary response protein MyD116 homolog, protein phosphatase 1 regulatory subunit 15A, protein phosphatase 1, regulatory (inhibitor) subunit 15A | |
Mouse | |
Affinity Purified | |
Apoptosis, Lipid and Metabolism, Unfolded Protein Response | |
23645.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction