Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD34 Mouse anti-Human, Mouse, Rat, Canine, Monkey, Allophycocyanin/Cy7, Clone: OTI3D12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271765APCCY7

Catalog No. N271765APC7


Add to cart

Description

Description

GADD34 Monoclonal antibody specifically detects GADD34 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications

Specifications

GADD34
OTI3D12
Flow Cytometry
Affinity Purified, Conjugated
PPP1R15A
Full length recombinant protein of human GADD34 (NP_055145) produced in HEK293T cells.
Immunogen affinity purified
Apoptosis, Lipid and Metabolism, Unfolded Protein Response
Primary
23645.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
GADD34Growth arrest and DNA damage-inducible protein GADD34, growth arrest and DNA-damage-inducible 34, Myeloid differentiation primary response protein MyD116 homolog, protein phosphatase 1 regulatory subunit 15A, protein phosphatase 1, regulatory (inhibitor) subunit 15A
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Canine, Human, Monkey, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit