Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GADD34 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GADD34
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093668
View Documents
Novus Biologicals
NBP303703100UL
100 μg
Each for $320.71
Add to cart
 
NB093669
View Documents
Novus Biologicals
NBP30370320UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

GADD34 Polyclonal antibody specifically detects GADD34 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

GADD34
Unconjugated
Rabbit
Apoptosis, Lipid and Metabolism, Unfolded Protein Response
PBS with 50% glycerol, pH7.3.
23645
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse
GADD34Growth arrest and DNA damage-inducible protein GADD34, growth arrest and DNA-damage-inducible 34, Myeloid differentiation primary response protein MyD116 homolog, protein phosphatase 1 regulatory subunit 15A, protein phosphatase 1, regulatory (inhibitor) subunit 15A
Recombinant fusion protein containing a sequence corresponding to amino acids 375-674 of human GADD34 (NP_055145.3).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit